accessibility menu, dialog, popup

UserName

CAS RN 913835-99-1

BNOHNCH3CH3H3CHOOH

CAS RN 913835-99-1

IUPAC Name: [5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid
Synonyms: 5-(tert-butylcarbamoyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C10H15BN2O3
InChi Key: FOJHBAIYLMKAIB-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=CN=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results