accessibility menu, dialog, popup

UserName

BOHNNCH3CH3HOHOHCl

CAS RN 913835-46-8

IUPAC Name: (4-{[2-(diethylamino)ethyl]carbamoyl}phenyl)boronic acid hydrochloride
Synonyms: 4-{[2-(diethylamino)ethyl]carbamoyl}phenylboronic acid hydrochloride
Molecular Weight (g/mol): 300.59
Molecular Formula: C13H22BClN2O3
InChi Key: WQYCSVUEHAFRHV-UHFFFAOYSA-N
SMILES: Cl.CCN(CC)CCNC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results