accessibility menu, dialog, popup

UserName

CAS RN 91366-65-3

NH2SOOFNON

CAS RN 91366-65-3

IUPAC Name: 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms: 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol): 217.17
Molecular Formula: C6H4FN3O3S
InChi Key: XROXHZMRDABMHS-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(F)C2=NON=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results