accessibility menu, dialog, popup

UserName

H3CNH3CONNNH

CAS RN 912761-62-7

IUPAC Name: N-ethyl-N-methyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Synonyms: N-ethyl-N-methyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 248.33
Molecular Formula: C13H20N4O
InChi Key: CMDQZBUDRBVXNM-UHFFFAOYSA-N
SMILES: CCN(C)C(=O)C1=C(N=CC=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results