accessibility menu, dialog, popup

UserName

CAS RN 91271-80-6

NH2N

CAS RN 91271-80-6

IUPAC Name: 1-{3-[(piperidin-1-yl)methyl]phenyl}methanamine
Synonyms: 1-[3-(piperidin-1-ylmethyl)phenyl]methanamine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: BMNFPARGJHNMAX-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(CN2CCCCC2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results