accessibility menu, dialog, popup

UserName

CAS RN 912569-78-9

CH3SHNH3CHCl

CAS RN 912569-78-9

IUPAC Name: methyl[(5-methylthiophen-2-yl)methyl]amine hydrochloride
Synonyms: methyl[(5-methylthiophen-2-yl)methyl]amine hydrochloride
Molecular Weight (g/mol): 177.69
Molecular Formula: C7H12ClNS
InChi Key: OQOUPFNJXCXICY-UHFFFAOYSA-N
SMILES: Cl.CNCC1=CC=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results