accessibility menu, dialog, popup

UserName

CAS RN 912350-01-7

BOHNFFFK

CAS RN 912350-01-7

IUPAC Name: potassium trifluoro[4-(phenylcarbamoyl)phenyl]boranuide
Synonyms: potassium trifluoro[4-(phenylcarbamoyl)phenyl]boranuide
Molecular Weight (g/mol): 303.13
Molecular Formula: C13H10BF3KNO
InChi Key: KWNGJSAKNIGTLH-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results