accessibility menu, dialog, popup

UserName

CAS RN 91059-97-1

CH3CH3NHNH2SH3C

CAS RN 91059-97-1

IUPAC Name: (2,4,6-trimethylphenyl)thiourea
Synonyms: 2,4,6-trimethylphenylthiourea
Molecular Weight (g/mol): 194.3
Molecular Formula: C10H14N2S
InChi Key: LBUGQLLKHAHCJJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC(N)=S)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results