accessibility menu, dialog, popup

UserName

CAS RN 910442-37-4

CH3OFFFHO

CAS RN 910442-37-4

IUPAC Name: 1-[2-(trifluoromethoxy)phenyl]ethan-1-ol
Synonyms: 1-[2-(trifluoromethoxy)phenyl]ethanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: LWOXLMRGAQEYMM-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results