CAS RN 91-78-1
IUPAC Name:
1,3,5-triphenyl-1,3,5-triazinane
Synonyms:
1,3,5-triphenyl-1,3,5-triazinane
Molecular Weight (g/mol):
315.42
Molecular Formula:
C21H21N3
InChi Key:
VASMRQAVWVVDPA-UHFFFAOYSA-N
SMILES:
C1N(CN(CN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Hexahydro-1,3,5-triphenyl-1,3,5-triazine, 98%, Thermo Scientific™
CAS: 91-78-1 Molecular Formula: C21H21N3 Molecular Weight (g/mol): 315.42 MDL Number: MFCD00043590 InChI Key: VASMRQAVWVVDPA-UHFFFAOYSA-N Synonym: hexahydro-1,3,5-triphenyl-1,3,5-triazine,1,3,5-triazine, hexahydro-1,3,5-triphenyl,anhydroformaldehyde aniline,1,3,5-triphenylhexahydro-1,3,5-triazine,1,3,5-triphenylhexahydro-s-triazine,unii-x3tz97444k,s-triazine, hexahydro-1,3,5-triphenyl,1,3,5-triphenyl-1,3,5-triazaperhydroine,anhydroformaldehyde aniline van,methyleneaniline trimer PubChem CID: 7065 IUPAC Name: 1,3,5-triphenyl-1,3,5-triazinane SMILES: C1N(CN(CN1C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4