accessibility menu, dialog, popup

UserName

CAS RN 91-15-6

NN

CAS RN 91-15-6

IUPAC Name: benzene-1,2-dicarbonitrile
Synonyms: phthalonitrile
Molecular Weight (g/mol): 128.13
Molecular Formula: C8H4N2
InChi Key: XQZYPMVTSDWCCE-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)