accessibility menu, dialog, popup

UserName

CAS RN 906352-98-5

CH3NNOOH

CAS RN 906352-98-5

IUPAC Name: {3-[(6-methylpyrazin-2-yl)oxy]phenyl}methanol
Synonyms: {3-[(6-methylpyrazin-2-yl)oxy]phenyl}methanol
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: DWEAUGVLWACZHO-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=CC(CO)=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results