accessibility menu, dialog, popup

UserName

CAS RN 906352-77-0

BOOH3CCH3H3CH3CFFFNO

CAS RN 906352-77-0

IUPAC Name: 4-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]morpholine
Synonyms: 4-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]morpholine
Molecular Weight (g/mol): 357.18
Molecular Formula: C17H23BF3NO3
InChi Key: FGPWVOFEKZVCDA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results