accessibility menu, dialog, popup

UserName

(R)(R)(S)O(R)(S)OOHOSOOOO(S)(S)(R)(R)O(R)OOSOOOOHn

CAS RN 9062-07-1

IUPAC Name: (1R,3R,4R,5S,8S)-8-{[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-2-yl]oxy}-3-{[(2S,3R,4R,5S,6R)-3-hydroxy-2-{[(1R,3S,4R,5S,8S)-3-hydroxy-4-(sulfonatooxy)-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-2,6-dioxabicyclo[3.2.1]octan-4-yl sulfate
Synonyms: carrageenan-iota eucheuma spinosum extract
Molecular Weight (g/mol): 946.75
Molecular Formula: C24H34O31S4
InChi Key: QIDSWKFAPCTSKL-RRQHLKGPSA-J
SMILES: OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](OS([O-])(=O)=O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]4[C@H]5CO[C@@H]4[C@@H](OS([O-])(=O)=O)[C@@H](O)O5)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O3)[C@H](O)[C@@H](O)[C@H]1OS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results