accessibility menu, dialog, popup

UserName

H2NOHO

CAS RN 90503-15-4

IUPAC Name: 1-amino-3-(benzyloxy)propan-2-ol
Synonyms: 1-amino-3-(benzyloxy)propan-2-ol
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: ZGHLNQMMKJNJIH-UHFFFAOYNA-N
SMILES: NCC(O)COCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results