accessibility menu, dialog, popup

UserName

A(a)

CAS RN 9050-97-9

IUPAC Name: tetramethylazanium chloride
Synonyms: tetramethylammonium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: OKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)C
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)