accessibility menu, dialog, popup

UserName

A(a)

CAS RN 9037-24-5

IUPAC Name: 1-bromo-2-methoxyethane
Synonyms: 2-bromoethyl methyl ether
Molecular Weight (g/mol): 138.99
Molecular Formula: C3H7BrO
InChi Key: YZUPZGFPHUVJKC-UHFFFAOYSA-N
SMILES: COCCBr
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Color 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)