accessibility menu, dialog, popup

UserName

CAS RN 90322-18-2

ONHN

CAS RN 90322-18-2

IUPAC Name: 3-{[(oxolan-2-yl)methyl]amino}propanenitrile
Synonyms: 3-[(oxolan-2-ylmethyl)amino]propanenitrile
Molecular Weight (g/mol): 154.21
Molecular Formula: C8H14N2O
InChi Key: ZSKZHSOMHSHFDF-UHFFFAOYNA-N
SMILES: N#CCCNCC1CCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)