accessibility menu, dialog, popup

UserName

CAS RN 902837-07-4

IUPAC Name: 1-(6-butoxypyridin-2-yl)piperazine
Synonyms: 1-(6-butoxypyridin-2-yl)piperazine
Molecular Weight (g/mol): 235.33
Molecular Formula: C13H21N3O
InChi Key: PPOHEYKESQFZEC-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=CC(=N1)N1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results