accessibility menu, dialog, popup

UserName

CH3NHOSOOH2N

CAS RN 902836-97-9

IUPAC Name: 4-{2-[(methylamino)methyl]phenoxy}benzene-1-sulfonamide
Synonyms: 4-{2-[(methylamino)methyl]phenoxy}benzenesulfonamide
Molecular Weight (g/mol): 292.35
Molecular Formula: C14H16N2O3S
InChi Key: AFXXBAMJRRMZDG-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1OC1=CC=C(C=C1)S(N)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results