accessibility menu, dialog, popup

UserName

CAS RN 902835-89-6

H3CH3CCH3OONHHNNI

CAS RN 902835-89-6

IUPAC Name: tert-butyl N-{2-[(3-iodopyridin-2-yl)amino]ethyl}carbamate
Synonyms: tert-butyl N-{2-[(3-iodopyridin-2-yl)amino]ethyl}carbamate
Molecular Weight (g/mol): 363.2
Molecular Formula: C12H18IN3O2
InChi Key: RHBATGHZRRPELS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCNC1=C(I)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results