accessibility menu, dialog, popup

UserName

H3COONHBrCl

CAS RN 902090-93-1

IUPAC Name: N-(4-bromo-2-chlorophenyl)-4-methoxybenzamide
Synonyms: N-(4-bromo-2-chlorophenyl)-4-methoxybenzamide
Molecular Weight (g/mol): 340.6
Molecular Formula: C14H11BrClNO2
InChi Key: QJZHFPOMZMQGEI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NC1=C(Cl)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results