accessibility menu, dialog, popup

UserName

Nn

CAS RN 9017-40-7

IUPAC Name: 1,2-diethenylbenzene; 4-ethenylpyridine
Synonyms: 4-vinylpyridine; divinylbenzene
Molecular Weight (g/mol): 235.33
Molecular Formula: C17H17N
InChi Key: HUEZUDXJCGHIHG-UHFFFAOYSA-N
SMILES: C=CC1=CC=NC=C1.C=CC1=CC=CC=C1C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)