accessibility menu, dialog, popup

UserName

H3COONH(R)(R)HO(S)O(S)O(R)OH(S)O(S)O(R)HO(S)O(S)O(R)OH(S)O(S)O(R)HO(S)O(S)O(R)OH(S)O(S)O(R)OH(S)HO(R)OH(R)NH2(R)HO(R)H2N(R)OH(R)NH2(R)HO(R)H2N(R)OH(R)NH2(R)HO(R)H2N(R)OHO(S)O(S)(R)HO(R)NH2(S)O(R)HOO(S)(R)HO(R)NH2(R)OHO(R)OH

CAS RN 9012-76-4

IUPAC Name: methyl N-[(2S,3R,4R,5S,6R)-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-5-{[(2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-{[(2R,3S,4R,5R,6S)-5-amino-6-{[(2R,3S,4R,5R,6R)-5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-6-(hydroxymethyl)oxan-3-yl]carbamate
Synonyms: chitosan
Molecular Weight (g/mol): 1526.46
Molecular Formula: C56H103N9O39
InChi Key: FLASNYPZGWUPSU-SICDJOISSA-N
SMILES: COC(=O)N[C@@H]1[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O[C@@H]5O[C@H](CO)[C@@H](O[C@@H]6O[C@H](CO)[C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7N)[C@H](O)[C@H]6N)[C@H](O)[C@H]5N)[C@H](O)[C@H]4N)[C@H](O)[C@H]3N)[C@H](O)[C@H]2N)[C@@H](CO)O[C@H]1O[C@H]1[C@H](O)[C@@H](N)[C@H](O[C@H]2[C@H](O)[C@@H](N)[C@H](O)O[C@@H]2CO)O[C@@H]1CO
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (2)
  • (9)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)