accessibility menu, dialog, popup

UserName

(R)(S)(R)O(R)OO(S)OO(R)O∗H3CO(R)O(R)CH3OH3CO(R)OOCH3OO∗H3COCH3On

CAS RN 9012-09-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C24H32O16)n
InChi Key: —
SMILES: CC(=O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)=O)OC(-*)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1O-*
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)