accessibility menu, dialog, popup

UserName

∗∗OOMeMen

CAS RN 9011-14-7

IUPAC Name: 1-methoxy-2-methyl-1-oxopropan-2-yl
Synonyms: poly(methyl methacrylate)
Molecular Weight (g/mol): 101.13
Molecular Formula: C5H9O2
InChi Key: PMAMJWJDBDSDHV-UHFFFAOYSA-N
SMILES: COC(=O)[C](C)C
Molecular Weight (g/mol) 
  • (3)
  • (11)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (11)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)