CAS RN 901-56-4
CAS RN 901-56-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
20(alpha)-Dihydro Pregnenolone, TRC
CAS: 901-56-4 Molecular Formula: C21 H34 O2 Molecular Weight (g/mol): 318.49 Synonym: Pregn-5-ene-3,20-diol, (3β,20S)-,Pregn-5-ene-3β,20α-diol (7CI,8CI),Pregnenediol (6CI),(3β,20S)-Pregn-5-ene-3,20-diol,20α-Dihydropregnenolone,20α-Dihydro pregnenolone,20α-Hydroxypregnenolone,3β,20α-Dihydroxypregn-5-ene,δ5-Pregnene-3β,20α-diol,δ5-Pregnenediol IUPAC Name: (3S,8S,9S,10R,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol SMILES: C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C