accessibility menu, dialog, popup

UserName

CAS RN 901-47-3

H3COONH2NNH2HNSOOCH3(E/Z)

CAS RN 901-47-3

IUPAC Name: methyl 2-amino-5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}pentanoate
Synonyms: methyl 2-amino-5-{[amino(4-methylbenzenesulfonamido)methylidene]amino}pentanoate
Molecular Weight (g/mol): 342.41
Molecular Formula: C14H22N4O4S
InChi Key: GIGVCLWIEMFWBF-UHFFFAOYNA-N
SMILES: COC(=O)C(N)CCCN=C(N)NS(=O)(=O)C1=CC=C(C)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results