accessibility menu, dialog, popup

UserName

HO(R)(S)O∗(R)HO(S)OH(S)OO∗OO∗(S)O(S)HOO(S)(R)(S)OHHOO∗BA

CAS RN 9005-32-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H8O7)A(C6H8O7)B
InChi Key: —
SMILES: O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)