accessibility menu, dialog, popup

UserName

CAS RN 9005-25-8

IUPAC Name: 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Synonyms: starch zea mays starch tapioca starch avena sativa starch 淀粉 solanum tuberosum starch pueraria lobata starch triticum vulgare starch oryza sativa starch
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Quantity 
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
Grade 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)