accessibility menu, dialog, popup

UserName

CAS RN 9004-74-4

OOHMen

CAS RN 9004-74-4

IUPAC Name: 2-methoxyethan-1-ol
Synonyms: 2-methoxyethanol MECS polyoxyethylene (40) monomethyl ether caswell no. 551 polyethylene glycol 300 methyl ether calcium salt methyl cellosolve EGME methylcelosolv methoxypolyethylene glycol polyethylene glycol 2000 monomethyl ether
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES: COCCO
Quantity 
  • (3)
  • (4)
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (8)
  • (8)
  • (3)
  • (4)
  • (2)
  • (7)
  • (3)