accessibility menu, dialog, popup

UserName

CAS RN 9004-65-3

H3COOOOOCH3HOOH3COHOCH3HOOCH3OHOCH3OHOCH3OHOH3COHOHCH3OOOOOH3COCH3OCH3OCH3OH3COH3COCH3

CAS RN 9004-65-3

IUPAC Name: 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Synonyms: 1-{[3,4,5-tris(2-hydroxypropoxy)-6-{[4,5,6-tris(2-hydroxypropoxy)-2-[(2-hydroxypropoxy)methyl]oxan-3-yl]oxy}oxan-2-yl]methoxy}propan-2-ol; methyl cellulose
Molecular Weight (g/mol): 1261.45
Molecular Formula: C56H108O30
InChi Key: PUSNGFYSTWMJSK-UHFFFAOYNA-N
SMILES: COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC.CC(O)COCC1OC(OC2C(COCC(C)O)OC(OCC(C)O)C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C(OCC(C)O)C1OCC(C)O
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (6)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)