accessibility menu, dialog, popup

UserName

Men

CAS RN 9003-07-0

IUPAC Name: 12-(3-octyloxiran-2-yl)dodecanoic acid
Synonyms: polypropylene
Molecular Weight (g/mol): 354.58
Molecular Formula: C22H42O3
InChi Key: NSYDMBURIUSUDH-UHFFFAOYNA-N
SMILES: CCCCCCCCC1OC1CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)