accessibility menu, dialog, popup

UserName

CH3OOOOHOHOOH3COHOH

CAS RN 9002-18-0

IUPAC Name: 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Synonyms: AGAR
Molecular Weight (g/mol): 336.34
Molecular Formula: C14H24O9
InChi Key: GYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILES: COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)