CAS RN 9000-65-1
CAS RN 9000-65-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
Tragacanth Gum MP Biomedicals
CAS: 9000-65-1 MDL Number: MFCD00131255 Synonym: Gum tragacanth