accessibility menu, dialog, popup

UserName

CAS RN 90-94-8

IUPAC Name: 4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
Synonyms: michler's ketone (michler's ketone)
Molecular Weight (g/mol): 268.36
Molecular Formula: C17H20N2O
InChi Key: VVBLNCFGVYUYGU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (1)