accessibility menu, dialog, popup

UserName

CAS RN 90-59-5

IUPAC Name: 3,5-dibromo-2-hydroxybenzaldehyde
Synonyms: 3,5-dibromsalicylaldehyd
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: JHZOXYGFQMROFJ-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Br)C=C1C=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (1)
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (3)
  • (5)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)