accessibility menu, dialog, popup

UserName

CAS RN 90-47-1

OO

CAS RN 90-47-1

IUPAC Name: 9H-xanthen-9-one
Synonyms: xanthone caswell no. 905 dibenzo-gamma-pyrone genicide benzophenone oxide diphenylene ketone oxide 9-xanthone epa pesticide chemical code 086503
Molecular Weight (g/mol): 196.21
Molecular Formula: C13H8O2
InChi Key: JNELGWHKGNBSMD-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2OC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)