accessibility menu, dialog, popup

UserName

CAS RN 90-00-6

H3CHO

CAS RN 90-00-6

IUPAC Name: 2-ethylphenol
Synonyms: 2-ethylphenol phlorol florol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: IXQGCWUGDFDQMF-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1O
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results