accessibility menu, dialog, popup

UserName

CAS RN 89929-93-1

OHO

CAS RN 89929-93-1

IUPAC Name: [3-(furan-2-yl)phenyl]methanol
Synonyms: [3-(furan-2-yl)phenyl]methanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: SBTRFADZILHXNG-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results