CAS RN 89929-82-8
CAS RN 89929-82-8
IUPAC Name:
[3-(thiophen-3-yl)phenyl]methanol
Synonyms:
[3-(thiophen-3-yl)phenyl]methanol
Molecular Weight (g/mol):
190.26
Molecular Formula:
C11H10OS
InChi Key:
VSWBZMMHNMDWJL-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC(=C1)C1=CSC=C1
1
–
1
of
1
results
1
(3-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™
CAS: 89929-82-8 Molecular Formula: C11H10OS Molecular Weight (g/mol): 190.26 MDL Number: MFCD06203083 InChI Key: VSWBZMMHNMDWJL-UHFFFAOYSA-N Synonym: 3-thien-3-ylphenyl methanol,3-thiophen-3-yl phenyl methanol,3-3-thienyl phenyl methanol,3-thiophen-3-ylphenyl methanol,acmc-20ls08,3-3-thienyl phenylmethanol,3-3-thiophenyl phenyl methanol,3-3-thienyl phenyl methan-1-ol PubChem CID: 7164662 IUPAC Name: (3-thiophen-3-ylphenyl)methanol SMILES: C1=CC(=CC(=C1)CO)C2=CSC=C2