accessibility menu, dialog, popup

UserName

CAS RN 898289-60-6

IUPAC Name: 2-{4-methyl-4H,6H,7H-thieno[3,2-c]pyran-4-yl}ethan-1-ol
Synonyms: 2-{4-methyl-6H,7H-thieno[3,2-c]pyran-4-yl}ethanol
Molecular Weight (g/mol): 198.28
Molecular Formula: C10H14O2S
InChi Key: UTWVZRFJFAVPIU-UHFFFAOYNA-N
SMILES: CC1(CCO)OCCC2=C1C=CS2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results