accessibility menu, dialog, popup

UserName

NNO

CAS RN 898289-23-1

IUPAC Name: 2-(pyrrolidin-1-yl)pyridine-4-carbaldehyde
Synonyms: 2-(pyrrolidin-1-yl)pyridine-4-carbaldehyde
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: QPKFMHRNVDJMLZ-UHFFFAOYSA-N
SMILES: O=CC1=CC(=NC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results