CAS RN 89691-88-3
CAS RN 89691-88-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5,7-Diazaspiro[3.4]octane-6,8-dione, TRC
CAS: 89691-88-3 Molecular Formula: C6H8N2O2 Molecular Weight (g/mol): 140.13 SMILES: O=C1NC(=O)C2(CCC2)N1