accessibility menu, dialog, popup

UserName

CAS RN 89581-38-4

CH3OONNBr

CAS RN 89581-38-4

IUPAC Name: methyl 5-bromopyrimidine-2-carboxylate
Synonyms: methyl 5-bromopyrimidine-2-carboxylate
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: XILAKTMDKMVJQV-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results