accessibility menu, dialog, popup

UserName

CAS RN 893749-85-4

IUPAC Name: 5-[(morpholin-4-yl)methyl]-1,2-oxazole-3-carbaldehyde
Synonyms: 5-(morpholin-4-ylmethyl)-1,2-oxazole-3-carbaldehyde
Molecular Weight (g/mol): 196.21
Molecular Formula: C9H12N2O3
InChi Key: AGLGFPIEVWZSLU-UHFFFAOYSA-N
SMILES: O=CC1=NOC(CN2CCOCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results