CAS RN 89267-41-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ethyl Methyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate (Photolysis Product), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Dehydronitrendipine, TRC
CAS: 89267-41-4 Molecular Formula: C18 H18 N2 O6 Molecular Weight (g/mol): 358.35 Synonym: Ethyl Methyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate,Nitrendipine Imp. A (EP),Photolysis Product of Nitrendipine IUPAC Name: 3-O-ethyl 5-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate SMILES: CCOC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2cccc(c2)[N+](=O)[O-]