accessibility menu, dialog, popup

UserName

CAS RN 892502-16-8

OONBr

CAS RN 892502-16-8

IUPAC Name: 2-bromo-6-(oxan-4-yloxy)pyridine
Synonyms: 2-bromo-6-(oxan-4-yloxy)pyridine
Molecular Weight (g/mol): 258.12
Molecular Formula: C10H12BrNO2
InChi Key: VLEKIKTZHYMZEW-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OC2CCOCC2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results