accessibility menu, dialog, popup

UserName

ONFFFCl

CAS RN 892502-15-7

IUPAC Name: 4-[2-(chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
Synonyms: 4-[2-(chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
Molecular Weight (g/mol): 279.69
Molecular Formula: C12H13ClF3NO
InChi Key: KCGVSWOLBMMCMW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(CCl)=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results