accessibility menu, dialog, popup

UserName

CAS RN 892502-02-2

CH3HNN

CAS RN 892502-02-2

IUPAC Name: methyl[(5-phenylpyridin-2-yl)methyl]amine
Synonyms: methyl[(5-phenylpyridin-2-yl)methyl]amine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: HHOWXRCPAWWJPH-UHFFFAOYSA-N
SMILES: CNCC1=NC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results